missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB5986RR-100UG
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
CCR1 Monoclonal specifically detects CCR1 in Mouse samples. It is validated for Flow Cytometry.
Tekniske data
| CCR1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| E. coli-derived recombinant mouse CCR1, Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281), Accession No. P51675 | |
| 100 μg | |
| 1230.0 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 643854R | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P51675 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects mouse CCR1 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion