missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR3 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB729G-100UG
This item is not returnable.
View return policy
Description
CCR3 Monoclonal specifically detects CCR3 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CCR3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
| Y3 rat myeloid cell line transfected with mouse CCR3 Met1-Phe359 Accession # P51678 | |
| 100 μg | |
| 1232 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 83101 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P51678 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse CCR3 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction