missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR5 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB18041R-100UG
This item is not returnable.
View return policy
Description
CCR5 Monoclonal specifically detects CCR5 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CCR5 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| C6 rat glioma cell line transfected with mouse CCR5, Accession No. P51682 | |
| 100 μg | |
| 1234.0 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 225307 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P51682 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse CCR5 transfectants but not the parental cell line in Flow Cytometry and Western blots. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction