missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD109 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7717
This item is not returnable.
View return policy
Description
CD109 Polyclonal specifically detects CD109 in Mouse samples. It is validated for Western Blot.
Specifications
| CD109 | |
| Polyclonal | |
| Unconjugated | |
| Q8R422 | |
| CD109 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD109 Ala22-Ser1269 Accession # Q8R422 | |
| 100 μg | |
| Primary | |
| Detects mouse CD109 in direct ELISAs and Western blots. In direct ELISAs, approximately 65% cross-reactivity with recombinant human CD109 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 135228 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction