Learn More
Mouse CD11b/Integrin alpha M Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems FAB11241T-100UG
Product Code. 18125391
  • € 463.00 / 100µg
Description
CD11b Monoclonal specifically detects CD11b in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD11b
Monoclonal
Alexa Fluor 594
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha M beta 2 Phe17-Asn1105 (Integrin alpha M), Gln24-Asn702 (Integrin beta 2) Accession # NP_032427 (Integrin alpha M) & P11835 (Integrin beta 2)
100 μg
3684
Mouse
Supernatant
Flow Cytometry
908649
Flow Cytometry 0.25-1μg/106 cells
NP_032427 (Integrin alpha M) & P11835 (Integrin beta 2)
Rat
Protein A or G purified from hybridoma culture supernatant
Primary
Detects recombinant mouse CD11b/Integrin alpha M in Western blot.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b