missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD177 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB8186U-100UG
This item is not returnable.
View return policy
Description
CD177 Monoclonal specifically detects CD177 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD177 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD177, CD177 antigenNB1 GP, CD177 molecule, HNA-2a, HNA2Acell surface receptor, Human neutrophil alloantigen 2a, NB1 GP, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1, PRV1NB1 glycoprotein | |
| Human embryonic kidney cell line HEK293-derived recombinant mouse CD177 Accession # Q8R2S8 | |
| RUO | |
| 57126 | |
| Mouse | |
| IgG |
| Flow Cytometry | |
| 1171A | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8R2S8 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Detects mouse CD177 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction