missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD25/IL-2R alpha Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2438
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Polyclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| CD25/IL-2R alpha | |
| Polyclonal | |
| Biotin | |
| Q544I2 | |
| IL2RA | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD25/IL-2 R alpha Glu22-Lys236 Accession # Q544I2 | |
| 50 μg | |
| Primary | |
| Detects mouse CD25/IL-2 R alpha in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) CD25/IL-2 R alpha is observed. In Western blots, approximately 5% cross-reactivity with rhCD25/IL-2 R alpha is observed and less than 1% cross-reactivity with recombinant mouse (rm) IL-2 R beta and rmIL-2 R gamma is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3559 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction