missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD30/TNFRSF8 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF852
This item is not returnable.
View return policy
Description
CD30/TNFRSF8 Polyclonal specifically detects CD30/TNFRSF8 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| CD30/TNFRSF8 | |
| Polyclonal | |
| Biotin | |
| Q60846 | |
| TNFRSF8 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD30/TNFRSF8 Phe19-Thr281 Accession # Q60846 | |
| 50 μg | |
| Primary | |
| Detects mouse CD30/TNFRSF8 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) CD30, recombinant mouse (rm) TNF RI, rhTNF RI, rmTNF RII, and rhTNF RII is observed. In Western blots, less than 1% cross-reactivity with rhCD30 is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 943 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction