missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD34 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB6518
This item is not returnable.
View return policy
Description
CD34 Monoclonal specifically detects CD34 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD34 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q64314 | |
| CD34 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD34 Thr35-Thr287 Accession # Q64314 | |
| 100 μg | |
| Primary | |
| Detects mouse CD34 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human, rat, or porcine CD34 is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 700011 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 947 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction