missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD36/SR-B3 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB25191
This item is not returnable.
View return policy
Description
CD36/SR-B3 Monoclonal specifically detects CD36/SR-B3 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| CD36/SR-B3 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| Q08857 | |
| CD36 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD36/SR-B3 Gly30-Lys439 Accession # Q08857 | |
| 100 μg | |
| Primary | |
| Detects mouse CD36/SR-B3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CD36 is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 324205 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BDPLT10, CD_antigen: CD36, CD36, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, Collagen R, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, PAS IV, PAS-4, PAS-4 protein, PASIV, Platelet collagen receptor, platelet glycoprotein 4, SCARB3, scavenger receptor class B, member 3, SRB3, SR-B3, Thrombospondin R, Thrombospondin receptor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 948 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction