missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD45 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB3507V-100UG
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD45 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_035340 | |
| PTPRC | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD45 isoform 3 Gln24-Lys425 Accession # NP_035340 | |
| RUO | |
| 5788 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 319211 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse CD45 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction