missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD45 Biotinylated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems BAM114
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Complement-dependent Cytotoxicity.
Specifications
| CD45 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation, Complement-dependent Cytotoxicity | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5788 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cytotoxicity Assay | |
| 30-F11 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| PTPRC | |
| Mouse thymus or spleen | |
| 250 μg | |
| Primary | |
| Detects mouse CD45. Detects both alloantigens and all isoforms of CD45. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction