missing translation for 'onlineSavingsMsg'
Learn More

Mouse CD68/SR-D1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Product Code. 18790587
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18790587 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18790587 Supplier R&D Systems Supplier No. IC101141R100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD68/SR-D1
Applications Flow Cytometry
Classification Monoclonal
Clone 2449D
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P31996
Gene Alias CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1, SRD1, SR-D1
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived mouse CD68, Asp21-Pro282, Accession # P31996
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 968
Test Specificity Detects mouse CD68 in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.