missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD74 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7478-SP
This item is not returnable.
View return policy
Description
CD74 Monoclonal specifically detects CD74 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD74 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P04441 | |
| CD74 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD74 Gln56-Cys215 Accession # P04441 | |
| 25 μg | |
| Primary | |
| Detects mouse CD74 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CD74 is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 829706 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD74, CD74 antigen, CD74 antigen (invariant polypeptide of major histocompatibility complex, classII antigen-associated), CD74 molecule, major histocompatibility complex, class II invariant chain, CLIP, DHLAG, DHLAGgamma chain of class II antigens, HLA class II histocompatibility antigen gamma chain, HLADG, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia antigen-associated invariant chain, Ia-associated invariant chain, Ia-GAMMA, Ii, INVG34, MHC HLA-DR gamma chain, p33 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 972 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction