missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD79B Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6814
This item is not returnable.
View return policy
Description
CD79B Polyclonal specifically detects CD79B in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| CD79B | |
| Polyclonal | |
| Unconjugated | |
| P15530 | |
| CD79B | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD79B Met29-Asp158 Accession # P15530 | |
| 100 μg | |
| Primary | |
| Detects mouse CD79B in direct ELISAs and Western blots. In direct ELISAs, approximately 55% of cross-reactivity with human CD79B is observed. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| B29, B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79B, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGB, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 974 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction