missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CDO Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF2429
This item is not returnable.
View return policy
Description
CDO Polyclonal specifically detects CDO in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CDO | |
| Polyclonal | |
| Biotin | |
| AAC43031 | |
| CDON | |
| Mouse myeloma cell line NS0-derived recombinant mouse CDO Val22-Leu963 Accession # AAC43031 | |
| 50 μg | |
| Primary | |
| Detects mouse CDO in Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 1.7-15 ug/mL | |
| CDON, Cdon homolog (mouse), CDOORCAM, cell adhesion molecule-related/down-regulated by oncogenes, MGC111524, ORCAM, surface glycoprotein, Ig superfamily member | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 50937 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction