missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CEACAM-1/CD66a Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB6480-SP
This item is not returnable.
View return policy
Description
CEACAM1/CD66a Monoclonal specifically detects CEACAM1/CD66a in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| CEACAM1/CD66a | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| P31809 | |
| Ceacam1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-1/CD66a Glu35-Gly428 Accession # P31809 | |
| 25 μg | |
| Primary | |
| Detects mouse CEACAM-1/CD66a in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CECAM-1, -3, -5, or -6 is observed. In Western blots, no cross-reactivity with rhCEACAM-1, -3, -4, -5, -6, or -7 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
| 723629 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antigen CD66, BGP1, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, Biliary Glycoprotein 1, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen, Cea-1, CEACAM1, Hv-1, Hv-2, MHVR | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 634 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction