missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CLEC9a Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB67762N-100UG
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal specifically detects CLEC9a in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CLEC9a | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD370, C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CLEC9a, Lys57-Ile264, Accession # EDK99924 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 2456A | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| EDK99924 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 283420 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction