missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cryptic Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB18401S-100UG
This item is not returnable.
View return policy
Description
CYRPTIC/CFC1 Monoclonal specifically detects CYRPTIC/CFC1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CYRPTIC/CFC1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P97766 | |
| CFC1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cryptic Tyr36-Gly177 Accession # P97766 | |
| RUO | |
| 55997 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 221428 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CFC1, CFC-1, CFC1B, cripto, FRL-1, cryptic family 1, Cryptic family protein 1, cryptic protein, CRYPTICFLJ77897, HTX2MGC133213 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Cryptic in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant human Cryptic is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction