missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Mouse Cxcl10 (P17515) Recombinant Protein

Product Code. 16121630
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
This item is not returnable. View return policy

Product Code. 16121630

Brand: Abnova™ P4609.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Sequence: IPLARTVRCNCIHIDDGPVRMRAIGKLEIIPASLSCPRVEIIATMKKNDEQRCLNPESKTIKNLMKAFSQKRSKRAP

Specifications

Accession Number P17515
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 15945
Molecular Weight (g/mol) 8.7kDa
Name Cxcl10 (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Quality Control Testing 1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
Quantity 25 μg
Immunogen IPLARTVRCNCIHIDDGPVRMRAIGKLEIIPASLSCPRVEIIATMKKNDEQRCLNPESKTIKNLMKAFSQKRSKRAP
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias C7/CRG-2/INP10/IP-10/IP10/Ifi10/Scyb10/gIP-10/mob-1
Common Name Cxcl10
Gene Symbol Cxcl10
Biological Activity The activity is determined by the ability to chemoattract primary human T cells and is detectable starting at 5ng/mL.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.