missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMX1A Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41193UV
This item is not returnable.
View return policy
Description
LMX1A Polyclonal antibody specifically detects LMX1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| LMX1A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 4009 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| LIM homeobox transcription factor 1, alpha, LMX1.1 | |
| Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human LMX1A. The immunogen is located within amino acids 300 - 350 of LMX1A. Amino Acid Squence: CPQMPGDHMHPYGAEPL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction