missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL4/PF4 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF595
This item is not returnable.
View return policy
Description
CXCL4/PF4 Polyclonal specifically detects CXCL4/PF4 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| CXCL4/PF4 | |
| Polyclonal | |
| Biotin | |
| Q9Z126 | |
| PF4 | |
| E. coli-derived recombinant mouse CXCL4/PF4 (R&D Systems, Catalog # 595-P4) Val30-Ser105 Accession # Q9Z126 | |
| 50 μg | |
| Primary | |
| Detects mouse CXCL4/PF4 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human (rh) CXCL4, recombinant mouse (rm) MIG, rmI-TAC, rhIL-8, and rhIP-10 is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5196 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction