missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL9/MIG Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems BAF492
This item is not returnable.
View return policy
Description
CXCL9/MIG Polyclonal specifically detects CXCL9/MIG in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| CXCL9/MIG | |
| Polyclonal | |
| Biotin | |
| P18340 | |
| CXCL9 | |
| E. coli-derived recombinant mouse CXCL9/MIG (R&D Systems, Catalog # 492-MM) Accession # P18340 | |
| 50 μg | |
| Primary | |
| Detects mouse CXCL9/MIG in ELISAs and Western blots. In sandwich immunoassays, less than 0.5% cross-reactivity with rhCXCL9 and 0.005% cross-reactivity with rmMIP-2, rrCINC-1, rhGRO alpha , rhGRO beta and rhGRO gamma is observed. | |
| Mouse | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4283 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction