missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCR2/IL-8RB Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB2164U-100UG
This item is not returnable.
View return policy
Description
CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CXCR2/IL-8RB | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P35343 | |
| CXCR2 | |
| C6 rat glioma cell line transfected with mouse CXCR2 Met1-Leu359 Accession # P35343 | |
| RUO | |
| 3579 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 242216 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD182, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, CMKAR2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 RB, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, IL8RB, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
| Rat | |
| 100 μg | |
| Primary | |
| Stains mouse CXCR2 transfected cells but not CXCR1 transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction