missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCR4 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB21651A-100UG
This item is not returnable.
View return policy
Description
CXCR4 Monoclonal specifically detects CXCR4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CXCR4 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and sodium azide | |
| CD184, chemokine (C-X-C motif) receptor 4, chemokine (C-X-C motif), receptor 4 (fusin), C-X-C chemokine receptor type 4, CXC-R4, CXCR-4, D2S201E, D2S201ESDF-1 receptor, FB22, Fusin, HM89, HM89NPYRL, HSY3RR, LAP3, LAP-3, LCR1, LESTR, LESTRCD184 antigen, Leukocyte-derived seven transmembrane domain receptor, leukocyte-derived seven-transmembrane-domain receptor, lipopolysaccharide-associated protein 3, neuropeptide Y receptor Y3, NPY3R, NPY3RWHIM, NPYR, NPYRL, NPYY3R, pCXCR4, seven transmembrane helix receptor, seven-transmembrane-segment receptor, spleen, Stromal cell-derived factor 1 receptor, WHIMS | |
| Rat | |
| 100 μg | |
| 7852 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 247506 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| P70658 | |
| CXCR4 | |
| Y3 rat myeloid cell line transfected with mouse CXCR4, Met1-Ser359, Accession No. P70658 | |
| Primary | |
| Detects mouse CXCR4 transfectants. Does not stain irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction