missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCR5 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB6198S-100UG
This item is not returnable.
View return policy
Description
CXCR5 Monoclonal specifically detects CXCR5 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CXCR5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q04683 | |
| CXCR5 | |
| E. coli-derived recombinant mouse CXCR5 Met1-Pro57 Accession # Q04683 | |
| RUO | |
| 643 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 614641 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| BLR1, BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXCR5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects recombinant mouse CXCR5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CXCR7 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction