missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DPPIV/CD26 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF954
This item is not returnable.
View return policy
Description
DPPIV/CD26 Polyclonal specifically detects DPPIV/CD26 in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| DPPIV/CD26 | |
| Polyclonal | |
| Unconjugated | |
| P28843 | |
| DPP4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse DPPIV/CD26 Ser29-His760 Accession # P28843 | |
| 100 μg | |
| Primary | |
| Detects mouse DPPIV/CD26 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant human DPPIV/CD26 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot .25 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1803 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction