missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EOMES Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC8889R-100UG
This item is not returnable.
View return policy
Description
EOMES Monoclonal specifically detects EOMES in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| EOMES | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR2, TBR-2, TBR2eomesodermin homolog, T-brain-2 | |
| E. coli-derived recombinant mouse EOMES Met1-Ser126 Accession # O54839 | |
| RUO | |
| 8320 | |
| Mouse | |
| IgG |
| Flow Cytometry | |
| 1219A | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O54839 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Detects mouse EOMES in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction