missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EPCR Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB2749U-100UG
This item is not returnable.
View return policy
Description
EPCR Monoclonal specifically detects EPCR in Mouse samples. It is validated for Flow Cytometry.
Specifications
| EPCR | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q64695 | |
| PROCR | |
| Mouse myeloma cell line NS0-derived recombinant mouse EPCR Ala17-Ser214 Accession # Q64695 | |
| RUO | |
| 10544 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 432714 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| APC receptor, CCCA, CCD41, CCD41centrosome-associated protein, CD201, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, PROCR, protein C receptor, endothelial | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse EPCR in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction