missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RII/RIII (CD32/CD16) Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF1460-SP
This item is not returnable.
View return policy
Description
Fc gamma RII/RIII (CD32/CD16) Polyclonal specifically detects Fc gamma RII/RIII (CD32/CD16) in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Fc gamma RII/RIII (CD32/CD16) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2214 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P08101 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIIB Thr40-Arg217 Accession # P08101 | |
| 25 μg | |
| Primary | |
| Detects mouse Fc gamma RII/RIII (CD32/CD16) in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Fc gamma RII is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction