missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FPRP/PTGFRN Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems BAF4495
This item is not returnable.
View return policy
Description
FPRP/PTGFRN Polyclonal specifically detects FPRP/PTGFRN in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| FPRP/PTGFRN | |
| Polyclonal | |
| Biotin | |
| NP_035327 | |
| Ptgfrn | |
| Mouse myeloma cell line NS0-derived recombinant mouse FPRP Arg22-Pro832 Accession # NP_035327 | |
| 50 μg | |
| Primary | |
| Detects mouse FPRP in Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
| CD315, CD9P-1, FPRP, PTGFRN | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5738 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction