missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse G-CSFR/CD114 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB60391S-100UG
This item is not returnable.
View return policy
Description
G-CSFR/CD114 Monoclonal specifically detects G-CSFR/CD114 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| G-CSFR/CD114 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P40223 | |
| CSF3R | |
| Mouse myeloma cell line NS0-derived recombinant mouse G-CSF R/CD114 Cys26-Asp626 Accession # P40223 | |
| RUO | |
| 1441 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 723806 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), CSF3R, Csfgr, G-CSF R, G-CSF receptor, GCSFR, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse G-CSF R/CD114 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) GM-CSF R alpha, rmM-CSF R, recombinant human (rh) G-CSF R alpha, or rhGM-CSF R beta is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction