missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Galectin-7 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB1304
This item is not returnable.
View return policy
Description
Galectin-7 Monoclonal specifically detects Galectin-7 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| Galectin-7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| AAK29385 | |
| LGALS7 | |
| E. coli-derived recombinant mouse Galectin-7 Ser2-Phe136 Accession # AAK29385 | |
| 500 μg | |
| Primary | |
| Detects mouse Galectin-7 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Galectin-1, -3, -4, recombinant human (rh) Galectin-2, -7, or -8 is observed. In Western blots, approximately 20% cross-reactivity with rhGalectin-3, approximately 10% cross-reactivity with rhGalectin-7, and no cross-reactivity with rhGalectin-1 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, ELISA | |
| 212923 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Gal-7, GAL7, galectin 7, galectin-7, Galectin7, HKL-14, lectin, galactoside-binding, soluble, 7, LGALS7, LGALS7A, p53-induced gene 1 protein, PI7, PIG1, TP53I1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3963 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction