missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GDF-6/BMP-13 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF855
This item is not returnable.
View return policy
Description
GDF-6/BMP-13 Polyclonal specifically detects GDF-6/BMP-13 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
| GDF-6/BMP-13 | |
| Polyclonal | |
| Unconjugated | |
| P43028 | |
| GDF6 | |
| E. coli-derived recombinant mouse GDF-6/BMP-13 Thr335-Arg454 Accession # P43028 | |
| 100 μg | |
| Primary | |
| Detects mouse GDF-6/BMP-13 in direct ELISAs. | |
| Mouse | |
| IgG |
| Immunohistochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL | |
| BMP13, BMP-13, CDMP2, GDF16, GDF6, GDF-6, growth differentiation factor 6, Growth/differentiation factor 16, growth/differentiation factor 6, KFS, KFS1, KFSL, MCOP4, MCOPCB6, MGC158100, MGC158101, SCDO4, SGM1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 392255 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction