missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Glycoprotein V/CD42d Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6990-SP
This item is not returnable.
View return policy
Description
Glycoprotein V/CD42d Polyclonal specifically detects Glycoprotein V/CD42d in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Glycoprotein V/CD42d | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| platelet glycoprotein V, CD42d, glycoprotein V (platelet), GP5, GPV | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2814 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| NP_032174 | |
| GP5 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Glycoprotein V/CD42d Gln17-Gly522 Accession # NP_032174 | |
| 25 μg | |
| Primary | |
| Detects mouse Glycoprotein V/CD42d in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction