missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GRIN1/NMDAR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
€ 201.00 - € 440.00
Specifications
| Antigen | NMDA R, NR1 Subunit |
|---|---|
| Clone | 1031915 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, Immunocytochemistry 8-25 ug/mL |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18701737
![]() |
R&D Systems
MAB10655-SP |
25 μg |
€ 201.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18780157
![]() |
R&D Systems
MAB10655-100 |
100 μg |
€ 440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NMDA R, NR1 Subunit Monoclonal antibody specifically detects NMDA R, NR1 Subunit in Mouse samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| NMDA R, NR1 Subunit | |
| Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 2902 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 1031915 | |
| Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| GluN1, MRD8, NDHMSD, NMDA R, NR1 Subunit, NMDA1, NMDAR1, NMD-R1, NR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse GRIN1/NMDAR1, Met1-Gln559 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title