missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Hematopoietic Lineage Marker Fluorescein Antibody Cocktail, R&D Systems™

Rat Monoclonal Antibody cocktail
Brand: R&D Systems FLC001F
Includes: 500μL of an AlexaFluor 700-conjugated antibody cocktail of the following mouse lineage markers:
- Anti-mouse CD3 (Clone 17A2)Anti-mouse B220 (Clone RA3-6B2)Anti-mouse CD11b (Clone M1/70)Anti-mouse GR-1 (Clone RB6-8C5)Anti-mouse Ter-119 (Clone TER119)Anti-mouse CD5 (Clone 53-7.3)
Description
Hematopoietic Lineage Marker Monoclonal specifically detects Hematopoietic Lineage Marker in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Hematopoietic Lineage Marker | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Unpurified | |
| RUO | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 17A2, RA3-6B2, M1/70, RB6-8C5, TER119, 53-7.3 | |
| Flow Cytometry | |
| Rat | |
| 25 Tests | |
| Primary | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction