missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFITM3/Fragilis Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF3377
This item is not returnable.
View return policy
Description
IFITM3/Fragilis Polyclonal specifically detects IFITM3/Fragilis in Mouse samples. It is validated for Western Blot.
Specifications
| IFITM3/Fragilis | |
| Polyclonal | |
| Biotin | |
| NP_079654 | |
| IFITM3 | |
| E. coli-derived recombinant mouse IFITM3/Fragilis Asn2-His57 Accession # NP_079654 | |
| 50 μg | |
| Primary | |
| Detects mouse IFITM3/Fragilis in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human IFITM3 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| 1-8U, Fragilis, interferon induced transmembrane protein 3 (1-8U), interferon-induced transmembrane protein 3, Interferon-inducible protein 1-8U, IP15, mil-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10410 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction