missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-alpha/beta R2 APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB1083A
This item is not returnable.
View return policy
Description
IFN-alpha/beta R2 Polyclonal specifically detects IFN-alpha/beta R2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IFN-alpha/beta R2 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFN-aR2, IFNAR2, IFNARB, IFNbR2, IFN-bR2, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3455 | |
| Mouse | |
| IgG |
| Flow Cytometry | |
| APC | |
| Q9D1R7 | |
| IFNAR2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IFN-alpha/beta R2 Ser22-Ala239 (Lys160Asn) Accession # Q9D1R7 | |
| 100 Tests | |
| Primary | |
| Detects mouse IFN-alpha/beta R2 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) IFN-gamma R2 and rmIL-20 R alpha is observed, 5% cross-reactivity with recombinant human (rh) IL-10 R beta , rmIFN-gamma RI, and rhIL-20 R beta is observed and less than 1% cross-reactivity with rhIFN-beta R alpha is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction