missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse IFN-gamma Platinum ELISA Kit

For the quantitative detection of mouse IFN-gamma.

Merk:  Invitrogen™ BMS606TWO

Overige gegevens : Gewicht : 0.23750kg

 Bekijk meerdere types van dit product via de

Produktcode. 15542017

  • € 632.00 / 2 stuks


Op voorraad 2
Toevoegen aan winkelmandje

Inclusief: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IFN-gamma
Biotin-Conjugate anti-mouse IFN-gamma monoclonal antibody
Streptavidin-HRP
Mouse IFN-gamma Standard lyophilized, 2ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Beschrijving

Beschrijving

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specificatie

Specificatie

P01580
Biotin
15978
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
2 x 96 tests
5.3pg/mL
50μL
Mouse
1 hr 20 min
IFN gamma
Immune interferon, IFG
5%
15.6-1,000 pg/mL
ELISA
Colorimetric Microplate Reader
ELISA
RUO
Cell culture supernatant, serum
Serum (50μL), Supernatants (50μL)
2-8°C
3 hr 10 min
Ifng
10%
HRP
Video's

For Research Use Only