missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-gamma R2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1185-SP
This item is not returnable.
View return policy
Description
IFN-gamma R2 Polyclonal specifically detects IFN-gamma R2 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| IFN-gamma R2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AF-1, AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFNgammaR2, IFN-gamma-R2, IFN-gR2, IFNGR2, IFNGT1, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3460 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.2-0.8 ug/mL | |
| NP_032364 | |
| IFNGR2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IFN-gamma R2 Ser21-Val243 Accession # NP_032364 | |
| 25 μg | |
| Primary | |
| Detects mouse IFN-gamma R2 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human IFN-gamma R2 is observed and less than 1% cross-reactivity with recombinant mouse (rm) IFN-gamma R1 and rmIFN-alpha/beta R2 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction