missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgE Antibody (LO-ME-3), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-42334AF532
This item is not returnable.
View return policy
Description
Mouse IgE Monoclonal antibody specifically detects Mouse IgE in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| Mouse IgE | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| LO-ME-3 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
| Immunoglobulin E, ImmunoglobulinE | |
| Mouse IgE | |
| 0.2 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction