missing translation for 'onlineSavingsMsg'
Learn More

Mouse IGF-I/IGF-1 Biotinylated Antibody, R&D Systems™

Product Code. 18146091
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18146091 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18146091 Supplier R&D Systems Supplier No. BAF791

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

IGF-I/IGF-1 Polyclonal specifically detects IGF-I/IGF-1 in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-I/IGF-1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL
Gene Accession No. Q8CAR0
Gene Alias IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C
Gene Symbols IGF1
Host Species Goat
Immunogen E. coli-derived recombinant mouse IGF-I/IGF-1 Gly33-Ala102 Accession # Q8CAR0
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3479
Test Specificity Detects mouse IGF-I/IGF-1 in ELISAs and Western blots. In sandwich immunoassays, less than 0.5% cross-reactivity with recombinant human IGF-I/IGF-1 is observed and less than 0.1% cross-reactivity with recombinant mouse IGF-II/IGF-2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.