Learn More
Mouse IgG3 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand:  R&D Systems IC007A
Product Code. 18134681
  • ONLINE EXCLUSIVE PRICE
    € 324.00
    € 305.55 / 200 tests
    Save € 18.45
    5% Off
Description
IgG3 Monoclonal specifically detects IgG3 in Mouse samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG3
133316
Control 10 uL/10^6 cells
DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809
Mouse
Protein A or G purified from hybridoma culture supernatant
3502
Mouse
IgG3
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
IGHG3
KLH
200 Tests
Selected for use as mouse IgG 3 isotype control for flow cytometry applications.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.