missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgG3 Mouse Uncoated ELISA Kit with Plates

Specifically engineered for accurate and precise measurement of mouse IgG3.

€ 242.00 - € 782.00

Specifications

Assay Range 1.875-125 ng/mL
Assay Sensitivity 1.875 ng/mL
Conjugate HRP
Product Type ELISA Kit
Sample Type Plasma, Serum, Supernatant
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15520897
View Documents Promotion Details
Invitrogen™
88-50440-22
2 x 96 Tests
€ 242.00
Pack of 2
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
 
15530897
View Documents Promotion Details
Invitrogen™
88-50440-76
20 x 96 Tests
€ 782.00
Pack of 20
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
 
15550897
View Documents Promotion Details
Invitrogen™
88-50440-86
10 x 96 Tests
€ 544.00
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 25-04-2024
to see stock.
Add to basket
 
Description

Description

The Mouse Immunoglobulin G3 (IgG3) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IgG3. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Mouse IgG3 Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

1.875-125 ng/mL
HRP
Plasma, Serum, Supernatant
ELISA
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86)
RUO
2°C to 8°C
Mouse
24 hr. 30 min.
1.875 ng/mL
ELISA Kit
Mouse
Colorimetric Microplate Reader
HRP
Plasma, 10 μL; Serum, 10 μL; Supernatant, 50 μL
Immunoglobulin G3
1 hr. 45 min.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only