missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG3 Rabbit anti-Mouse, Clone: RM218 (Biotin), Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB14939.50ug
This item is not returnable.
View return policy
Description
Rabbit monoclonal antibody raised against native mouse IgG3.
- High specificity
- High reactivity
Specifications
| Mouse IgG3 | |
| Monoclonal | |
| Biotin | |
| ELISA (0.005-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blotting (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| P03987 | |
| AI324046 | |
| Native purified mouse IgG3. | |
| 50 μg | |
| Primary | |
| Mouse | |
| Liquid |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| RM218 | |
| Rabbit monoclonal antibody raised against native mouse IgG3. | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| IGHG3/IgG3/MGC68300 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 380795 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction