missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-1 alpha/IL-1F1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 14 publications
Brand: R&D Systems AF-400-SP
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Polyclonal specifically detects IL-1 alpha/IL-1F1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications
| IL-1 alpha/IL-1F1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3552 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL, Neutralization 0.002-0.004 ug/mL | |
| Q62161 | |
| IL1A | |
| E. coli-derived recombinant mouse IL-1 alpha/IL-1F1 Ser6-Ser161 Accession # Q62161 | |
| 25 μg | |
| Primary | |
| Detects mouse IL-1 alpha/IL-1F1 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant rat IL-1 alpha and recombinant cotton rat IL-1 alpha is observed and less than 1% cross-reactivity with recombinant human IL-1 alpha is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction