missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse FGL2 HA-tag Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Mouse Fc gamma RIIB/CD32b (Catalog # 1460-CD ) is immobilized at 1 μg/mL (100 μL/well), Recombinant Mouse FGL2 HA-tag (Catalog # 10691-FL) binds with an ED50 of 0.1-0.8 μg/mL. Measured by its ability to inhibit anti-CD3-induced proliferation of mouse T lymphocytes. The ED50 for this effect is 1.5-12 μg/mL.
Specifications
Specifications
| Accession Number | P12804.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 10875 |
| Molecular Weight (g/mol) | M.W.-observed: 61-72 kDa, under reducing conditions, M.W.-theroretical: 48 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse FGL2 protein Hemagglutinin Tag (YPYDVPDYA) Mouse FGL2 (Val20-Pro432) (c-terminus) Accession # P12804.1 N-terminus C-terminμs |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | fibrinogen-like 2, Fibrinogen-like protein 2, fibroleukin, pT49T49, T49 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction