missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-15 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems BAF447
This item is not returnable.
View return policy
Description
IL-15 Polyclonal specifically detects IL-15 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Detection (Matched Antibody Pair).
Specifications
| IL-15 | |
| Polyclonal | |
| Biotin | |
| P48346 | |
| IL15 | |
| E. coli-derived recombinant mouse IL-15 Asn49-Ser162 Accession # P48346 | |
| 50 μg | |
| Primary | |
| Detects mouse IL-15 in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant human IL-15 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| IL15, IL-15MGC9721, interleukin 15, interleukin-15 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3600 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction