missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-15R alpha APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB5511A-100
This item is not returnable.
View return policy
Description
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IL-15R alpha | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD215, CD215 antigen, IL15 R alpha, IL-15 R alpha, IL-15 receptor subunit alpha, IL15RA, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3601 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 888220 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q60819 | |
| IL15RA | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-15 R alpha Gly33-Lys205 Accession # Q60819 | |
| 100 Tests | |
| Primary | |
| Detects mouse IL-15 R alpha in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction